Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKBP6 Antibody, Alexa Fluor™ 488, Novus Biologicals™
SDP

Catalog No. NB148588 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB148588 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB148588 Supplier Novus Biologicals Supplier No. NBP297729AF488
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FKBP6 Polyclonal specifically detects FKBP6 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FKBP6
Applications Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 488
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias 36 kDa FKBP, EC 5.2.1.8, FK506 binding protein 6, 36kDa, FK506-binding protein 6, FK506-binding protein 6 (36kD), FKBP-36, FKBP36MGC87179, FKBP-6, Immunophilin FKBP36, peptidyl-prolyl cis-trans isomerase FKBP6,36 kDa FK506-binding protein, PPIase, PPIase FKBP6, rotamase
Host Species Rabbit
Immunogen Produced in rabbits immunized with E. coli-derived Human FKBP6 fragment.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8468
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.