Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ FKBP7 Polyclonal Antibody
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | FKBP7 |
---|---|
Concentration | 1 mg/mL |
Content And Storage | -20°C |
Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
Classification | Polyclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5119120
|
Invitrogen™
PA5119120 |
200 μL |
Each for $511.50
|
|
|||||
PIPA5121983
|
Invitrogen™
PA5121983 |
20 μL |
Each for $209.50
|
|
|||||
Description
FKBP7 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P)
The protein encoded by this gene belongs to the FKBP-type peptidyl-prolyl cis/trans isomerase (PPIase) family. Members of this family exhibit PPIase activity and function as molecular chaperones. A similar protein in mouse is located in the endoplasmic reticulum and binds calcium.Specifications
FKBP7 | |
-20°C | |
Polyclonal | |
Liquid | |
IgG | |
Human | |
FKBP7 | |
23 kDa FK506-binding protein; 23 kDa FKBP; 23kDa; FK506 binding protein 7; FK506-binding protein 23; FK506-binding protein 7; FKBP23; FKBP-23; Fkbp7; FKBP-7; Peptidyl-prolyl cis-trans isomerase FKBP7; PPIase; PPIase FKBP7; rotamase; UNQ670/PRO1304 | |
FKBP7 | |
Primary | |
Antigen affinity chromatography |
1 mg/mL | |
Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
Unconjugated | |
Rabbit | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9Y680 | |
51661 | |
Recombinant protein FK506 Binding Protein 7. The antigen corresponds to amino acid range 24-217 of the target protein. | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title