Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKRP Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen FKRP
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP16902920
SDP
View Documents
Novus Biologicals
NBP16902920UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP169029
SDP
View Documents
Novus Biologicals
NBP169029
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

FKRP Polyclonal specifically detects FKRP in Mouse samples. It is validated for Western Blot.
Specifications

Specifications

FKRP
Polyclonal
Rabbit
Q8CG64
79147
Synthetic peptides corresponding to Fkrp (fukutin related protein) The peptide sequence was selected from the C terminal of Fkrp. Peptide sequence LDHRQDVEFPEHFLQPLVPLPFAGFMAQAPNNYRRFLELKFGPGVIENPE.
Primary
Western Blot
Unconjugated
RUO
EC 2.-, fukutin related protein, fukutin-related protein, LGMD2IFLJ12576, MDC1CMGC2991, MDDGA5, MDDGB5, MDDGC5
FKRP
IgG
55 kDa
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.