Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FKTN Rabbit anti-Human, Mouse, Rat, Clone: 7J5N6, Novus Biologicals™
SDP

Catalog No. NB166040 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB166040 100 μg
NB166041 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. NB166040 Supplier Novus Biologicals Supplier No. NBP315472100UL
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

FKTN Monoclonal antibody specifically detects FKTN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)

Specifications

Antigen FKTN
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 7J5N6
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias CMD1X, EC 2.-, FCMDMGC134945, fukutin, Fukuyama type congenital muscular dystrophy (fukutin), Fukuyama type congenital muscular dystrophy protein, Fukuyama-type congenital muscular dystrophy protein, LGMD2MMGC134944, MDDGA4, MDDGB4, MDDGC4, MGC126857, MGC138243
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 362-461 of human FKTN (O75072).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) 2218
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.