Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flagellin Antibody (06), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB327682 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB327682 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB327682 Supplier Novus Biologicals Supplier No. NBP305967APC
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Flagellin Monoclonal antibody specifically detects Flagellin in Bacteria samples. It is validated for Western Blot, ELISA

Specifications

Antigen Flagellin
Applications Western Blot, ELISA
Classification Monoclonal
Clone 06
Conjugate APC
Dilution Western Blot, ELISA
Formulation PBS
Host Species Mouse
Immunogen This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Listeria monocytogenes Flagellin (Accession#: YP_013330.1; Met1-Ser287).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Bacteria
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.