Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flagellin Antibody (121), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NB072397 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072397 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072397 Supplier Novus Biologicals Supplier No. NBP290335AF532
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Flagellin Monoclonal specifically detects Flagellin in Bacteria samples. It is validated for ELISA.

Specifications

Antigen Flagellin
Applications ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 121
Conjugate Alexa Fluor 532
Dilution ELISA
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Listeria monocytogenes Flagellin (YP_013330.1; Met1-Ser287).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Listeria monocytogenes Listeria monocytogenes flagellin/FlaA
Target Species Bacteria
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.