Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flamingo homolog 1/CELSR3 Antibody (763103), HRP, Novus Biologicals™
SDP

Catalog No. NB374887 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB374887 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB374887 Supplier Novus Biologicals Supplier No. FAB7278H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Flamingo homolog 1/CELSR3 Monoclonal antibody specifically detects Flamingo homolog 1/CELSR3 in Human samples. It is validated for Western Blot, Flow Cytometry, CyTOF

Specifications

Antigen Flamingo homolog 1/CELSR3
Applications Western Blot, Flow Cytometry, CyTOF
Classification Monoclonal
Clone 763103
Conjugate HRP
Dilution Western Blot, Flow Cytometry, CyTOF-ready
Formulation PBS
Gene Alias Cadherin family member 11, cadherin, EGF LAG seven-pass G-type receptor 3 (flamingo homolog, Drosophila), CDHF11RESDA1, EGFL1cadherin EGF LAG seven-pass G-type receptor 3, EGF-like protein 1, EGF-like-domain, multiple 1, epidermal growth factor-like 1, Epidermal growth factor-like protein 1, Flamingo homolog 1, FMI1anchor protein, HFMI1, KIAA0812, MEGF2cadherin EGF LAG seven-pass G-type receptor 3, flamingo (Drosophila) homolog, multiple EGF-like domains 2, Multiple EGF-like domains protein 2, Multiple epidermal growth factor-like domains protein 2
Host Species Mouse
Immunogen E. coli-derived recombinant human CELSR3, Val531-Ser711, Accession # Q9NYQ7
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline GPCR
Primary or Secondary Primary
Gene ID (Entrez) 1951
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.