Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAP Antibody (SR1238), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP32225150UL
Description
FLAP Monoclonal antibody specifically detects FLAP in Human samples. It is validated for Western BlotSpecifications
FLAP | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Western Blot | |
SR1238 | |
Western Blot 1:500-1:2000 | |
arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein | |
A synthesized peptide derived from human FLAP (Uniprot #: P20292) | |
50 μg | |
Immunology, Inflammation | |
241 | |
Store at -20° C. Avoid freeze/thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction