Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FLAP Antibody (SR1238), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | FLAP |
---|---|
Clone | SR1238 |
Dilution | Western Blot 1:500-1:2000 |
Applications | Western Blot |
Classification | Monoclonal |
Description
FLAP Monoclonal antibody specifically detects FLAP in Human samples. It is validated for Western BlotSpecifications
FLAP | |
Western Blot 1:500-1:2000 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
arachidonate 5-lipoxygenase-activating protein, FLAPMK-886-binding protein | |
A synthesized peptide derived from human FLAP (Uniprot #: P20292) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR1238 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Immunology, Inflammation | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
241 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title