Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLASH Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB362166 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362166 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362166 Supplier Novus Biologicals Supplier No. NB500218F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLASH Polyclonal antibody specifically detects FLASH in Human, Mouse samples. It is validated for Western Blot

Specifications

Antigen FLASH
Applications Western Blot
Classification Polyclonal
Conjugate FITC
Dilution Western Blot
Formulation PBS
Gene Alias CASP8 associated protein 2, caspase 8 associated protein 2, CED-4, FLASHKIAA1315CASP8-associated protein 2, FLICE associated huge, FLICE-associated huge protein, FLJ11208, human FLASH, RIP25FLASH homolog RIP25
Host Species Rabbit
Immunogen N-terminus of the FLASH protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 9994
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.