Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Flat Cap Tubes PROMO
Click to view available options
:
Thermo Scientific™ Flat Cap Tubes
Description
0.2 mL individual tubes with integral snap shut caps are suitable for 0.2 mL thermal cycler blocks. They have a 0.25 mL maximum volume when closed. These individual tubes feature thin-wall design and are made from virgin polypropylene. The tubes are cleanroom produced, ensuring that they are free from contaminants that could affect experimental results. They are available in multiple colors with flat or domed caps.
Highlights
- Suitable for 0.2 mL thermal cycler blocks
- 0.25 mL maximum tube capacity when closed
- Integral snap shut cap
- Available with flat or domed caps
Applications
- PCR
- Sample storage
Additional Information
- Hazardous: No
- Storage Condition: Ambient
- PCR; Sample storage
Specifications
Specifications
Certifications/Compliance | Certified DNA/RNase Free |
Color | Natural |
Volume (Metric) Working | 0.2 mL |
Volume (Metric) | 0.25 mL |
Quantity | 1000 Tubes |
For Use With (Equipment) | Thermal Cyclers |
Material | Polypropylene |
Reaction Speed | Standard |
Closure Type | Flat Cap |
Format | 1,000 Tubes |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction