Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flavivirus Group Antigen Mouse anti-Virus, Clone: D1-4G2-4-15, MilliporeSigma™
Mouse Monoclonal Antibody
$206.00 - $517.50
Specifications
Antigen | Flavivirus Group Antigen |
---|---|
Clone | D1-4G2-4-15 |
Concentration | 1.0 mg/mL |
Applications | Immunofluorescence |
Classification | Monoclonal |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MAB10216I25
|
MilliporeSigma
MAB10216I25UG |
25 μg |
Each for $206.00
|
|
|||||
MAB10216I10
|
MilliporeSigma
MAB10216I100UG |
100 μg |
Each for $517.50
|
|
|||||
Description
Flavivirus Group Antigen Monoclonal antibody specifically detects Flavivirus Group Antigen in Virus samples. It is validated for Immunofluorescence.Specifications
Flavivirus Group Antigen | |
1.0 mg/mL | |
Monoclonal | |
Mouse | |
Virus | |
Dengue virus type 2 antigens. | |
Primary | |
Stable for 1 year at +2°C to +8°C from date of receipt. |
D1-4G2-4-15 | |
Immunofluorescence | |
Unconjugated | |
RUO | |
Purified mouse monoclonal antibody IgG2a in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
IgG2a | |
Affinity Purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title