Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLI1 Antibody (FLI1/3183), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB385315 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB385315 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB385315 Supplier Novus Biologicals Supplier No. NBP308652G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FLI1 Monoclonal antibody specifically detects FLI1 in Human samples. It is validated for ELISA, Protein Array

Specifications

Antigen FLI1
Applications ELISA, Protein Array
Classification Monoclonal
Clone FLI1/3183
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias EWSR2, Friend leukemia integration 1 transcription factor, Friend leukemia virus integration 1, Proto-oncogene Fli-1, SIC-1, Transcription factor ERGB
Host Species Mouse
Immunogen Recombinant fragment (aa 107-261) of human FLI1 protein with hexa-histidine tag (Uniprot: Q01543 )
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Cancer, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 2313
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.