Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLIP Antibody (NF6), FITC, Novus Biologicals™
SDP

Catalog No. NB173096 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB173096 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB173096 Supplier Novus Biologicals Supplier No. NBP280081F
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

FLIP Monoclonal antibody specifically detects FLIP in Human samples. It is validated for Immunohistochemistry, Immunofluorescence

Specifications

Antigen FLIP
Applications Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone NF6
Conjugate FITC
Formulation PBS
Gene Alias CASHFADD-like antiapoptotic molecule 1, CASP8 and FADD-like apoptosis regulator, CASP8AP1, Caspase homolog, Caspase-eight-related protein, Caspase-like apoptosis regulatory protein, caspase-related inducer of apoptosis, CASPER, CasperFLAME1, Cellular FLICE-like inhibitory protein, c-FLIPR, c-FLIPS, CLARPMACH-related inducer of toxicity, FADD-like apoptotic molecule, FLAME, FLAME-1, I-FLICEMRITc-FLIPFLIPc-FLIPL, Inhibitor of FLICE, Usurpin, usurpin beta
Host Species Mouse
Immunogen Recombinant human FLIP (aa 1-480).
Purification Method >95%, by SDS-PAGE
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Cancer, Cell Biology, Inhibitors Activators and Regulators, Signal Transduction, Tumor Suppressors
Primary or Secondary Primary
Gene ID (Entrez) 8837
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.