Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ10808 Antibody, Biotin, Novus Biologicals™
SDP

Catalog No. NB005269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB005269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB005269 Supplier Novus Biologicals Supplier No. NBP224534B
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLJ10808 Polyclonal specifically detects FLJ10808 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FLJ10808
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Biotin
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation PBS with 0.05% Sodium Azide
Gene Alias E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6
Gene Symbols UBA6
Host Species Rabbit
Immunogen A portion of amino acids 775-825 of human UBE1L2 was used as the immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.