Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ10808 Antibody, Novus Biologicals™
SDP

Catalog No. NBP224587SS Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP224587SS 0.025 mg
NBP224587 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP224587SS Supplier Novus Biologicals Supplier No. NBP224587SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLJ10808 Polyclonal specifically detects FLJ10808 in Human, Primate samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FLJ10808
Applications Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-3 ug/ml, Immunohistochemistry-Paraffin 10 ug/ml
Formulation 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide
Gene Accession No. A0AVT1
Gene Alias E1-L2, FLJ10808, FLJ23367, Monocyte protein 4, MOP4, MOP-4, UBA6, ubiquitin-activating enzyme E1, UBE1L2, Ubiquitin-activating enzyme 6, ubiquitin-activating enzyme E1-like 2, Ubiquitin-activating enzyme E1-like protein 2, ubiquitin-like modifier activating enzyme 6, ubiquitin-like modifier-activating enzyme 6
Gene Symbols UBA6
Host Species Rabbit
Immunogen A portion of amino acids 300-350 of human UBA6 was used as the immunogen for this antibody.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55236
Test Specificity The amino acid sequence used as immunogen for this antibody is 100% homologous in human (isoforms CRA_a and CRA_b) and chimpanzee, and 93% homologous in monkey.
Target Species Human, Primate
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.