Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ13946 Antibody, Novus Biologicals™
SDP

Catalog No. NB396322 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB396322 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB396322 Supplier Novus Biologicals Supplier No. NBP24679225UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLJ13946 Polyclonal antibody specifically detects FLJ13946 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen FLJ13946
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/ Immunofluorescence 0.25-2 μg/mL, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS (pH 7.2), 40% Glycerol
Gene Accession No. Q8N4P2
Gene Alias FLJ13946, FLJ77601, IFT70A, tetratricopeptide repeat domain 30A, tetratricopeptide repeat protein 30A, TPR repeat protein 30A
Host Species Rabbit
Immunogen This antibody was developed against Recombinant Protein corresponding to amino acids: FRKSVEFCNDHDVWKLNVAHVLFMQENKYKEAIGFYEPIVKKHYDNILNVSAIVLANLCVSYIMTSQNEEAEELMRKIEKEEEQLSYDDP
Purification Method Immunogen affinity purified
Quantity 25 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 92104
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.