Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLJ21908 Rabbit anti-Human, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB024683 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB024683 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB024683 Supplier Novus Biologicals Supplier No. NBP295263

Rabbit Polyclonal Antibody

FLJ21908 Polyclonal specifically detects FLJ21908 in Human samples. It is validated for Western Blot.

Specifications

Antigen FLJ21908
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:500 - 1:2000
Formulation PBS, 50% Glycerol
Gene Alias FLJ21908, RNA polymerase II associated protein 3, RNA polymerase II-associated protein 3, spag
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 17-241 of human RPAP3 (NP_001139547).
Purification Method IgG purified
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79657
Target Species Human
Content And Storage Store at -20°C.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.