Learn More
Description
Specifications
Specifications
| Antigen | FLNC |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 1.0 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/ml, Flow Cytometry 1 ug/million cells/ml, Immunohistochemistry 1:200, Immunocytochemistry/Immunofluorescence 2 ug/ml, Immunohistochemistry-Paraffin 1:200 |
| Formulation | PBS with 0.02% Sodium Azide |
| Gene Alias | ABP-280, ABP280A, ABP-280-like protein, ABPA, ABP-L, ABPLABP-L, gamma filamin, Actin-binding-like protein, filamin 2, filamin C, gamma, filamin C, gamma (actin binding protein 280), Filamin-2, filamin-C, FLJ10186, FLN2actin binding protein 280, FLNc, FLN-C, Gamma-filamin |
| Gene Symbols | FLNC |
| Host Species | Rabbit |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
