Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLNC Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB010655 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB010655 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB010655 Supplier Novus Biologicals Supplier No. NBP279816C
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLNC Polyclonal specifically detects FLNC in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen FLNC
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias ABP-280, ABP280A, ABP-280-like protein, ABPA, ABP-L, ABPLABP-L, gamma filamin, Actin-binding-like protein, filamin 2, filamin C, gamma, filamin C, gamma (actin binding protein 280), Filamin-2, filamin-C, FLJ10186, FLN2actin binding protein 280, FLNc, FLN-C, Gamma-filamin
Gene Symbols FLNC
Host Species Rabbit
Immunogen Recombinant protein made to an internal portion of the human FLNC protein (amino acids 270-468) [UniProt Q14315]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.