Learn More
FLNC Rabbit, anti-Human, Unconjugated, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | FLNC |
---|---|
Concentration | 300 μg/mL |
Applications | Western Blot, ELISA |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-223-8713
![]() ![]() |
Proteintech Group Inc
284921AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Filamin C (FLNC; also known as γ-FLN; ABP-L; FLN2), a muscle-specific filamin and a large actin-cross-linking protein. Human filamins are ∼280 kDa homodimers, each monomer consisting of an N-terminal actin-binding domain followed by 24 immunoglobulin (Ig)-like domains (d1 to d24), the last of which mediates dimerization. FLNC is specifically expressed in cardiomyocytes and skeletal myocytes and is involved in the maintenance of structural integrity. High filamin C associated with better prognosis of prostate cancer, leukemia and breast cancer patients. ( PMID: 25577646, PMID: 30867563, PMID: 31131323)
Specifications
FLNC | |
Western Blot, ELISA | |
Unconjugated | |
Rabbit | |
2318 |
300 μg/mL | |
Polyclonal | |
Liquid | |
Human | |
Primary |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.