Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flotillin 2 Antibody, FITC, Novus Biologicals™
SDP

Catalog No. NB054989 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB054989 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB054989 Supplier Novus Biologicals Supplier No. NBP298723F
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Flotillin 2 Polyclonal specifically detects Flotillin 2 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Flotillin 2
Applications Immunofluorescence
Classification Polyclonal
Conjugate FITC
Dilution Immunocytochemistry/Immunofluorescence
Formulation PBS
Gene Alias Epidermal surface antigen, ESA1Membrane component chromosome 17 surface marker 1, ESAchromosome 17, surface marker 1 (35kD protein identified bymonoclonal ECS-1), flotillin 2
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of human Flotillin 2.
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2319
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.