Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flotillin 2 Antibody (SR1814), Novus Biologicals™
SDP

Catalog No. NB260005 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB260005 50 μg
NB259608 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB260005 Supplier Novus Biologicals Supplier No. NBP32161350UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Flotillin 2 Monoclonal antibody specifically detects Flotillin 2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen Flotillin 2
Applications Western Blot, Immunoprecipitation
Classification Monoclonal
Clone SR1814
Conjugate Unconjugated
Dilution Western Blot 1:500-1:2000, Immunoprecipitation 1:30
Formulation PBS, pH 7.4, 150mM NaCl, 50% glycerol.
Gene Alias Epidermal surface antigen, ESA1Membrane component chromosome 17 surface marker 1, ESAchromosome 17, surface marker 1 (35kD protein identified bymonoclonal ECS-1), flotillin 2
Host Species Rabbit
Immunogen A synthesized peptide derived from human Flotillin 2 (Uniprot #: Q14254)
Purification Method Affinity purified
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2319
Target Species Human, Mouse, Rat
Content And Storage Store at -20° C. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.