Learn More
Johannes Lieder Flowers and Fruits of Rosaceae
Shop All Johannes Lieder GmbH & Co. KG Products
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Description
- 6100 Flowers and Fruits of Rosaceae
- 12 Microscope Slides
- Cherry (Prunus avium), flower bud t.s.
- Cherry, flower bud l.s. showing perigynous ovary
- Cherry, young stony fruit l.s.
- Apple (Pyrus malus), flower l.s. showing hypogynous ovary
- Apple, young pome t.s., a fleshy many seeded fruit
- Apple, young pome l.s.
- Gooseberry (Ribes uva-crispa), flower l.s.
- Gooseberry, many seeded berry l.s.
- Raspberry (Rubus idaeus), flower showing many carpels l.s.
- Raspberry, young aggregate fruit l.s.
- Strawberry (Fragaria), flower l.s.
- Strawberry, young aggregate fruit l.s.
Specifications
Specifications
| Classification | Botany |
| Material | Elastin Stained Glass |
| Mounting Type | Cross and Longitudinal Section |
| Type | Microscope Slides |
| Slide Type | Prepared Slide |