Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLRT1 Antibody, DyLight 488, Novus Biologicals™
SDP

Catalog No. NB089400 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB089400 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB089400 Supplier Novus Biologicals Supplier No. NBP299785G
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLRT1 Polyclonal specifically detects FLRT1 in Human samples. It is validated for ELISA.

Specifications

Antigen FLRT1
Applications ELISA
Classification Polyclonal
Conjugate DyLight 488
Formulation 50 mM Sodium Borate
Gene Alias fibronectin leucine rich transmembrane protein 1, Fibronectin-like domain-containing leucine-rich transmembrane protein 1, leucine-rich repeat transmembrane protein FLRT1, MGC21624
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FLRT1 (Q9NZU1-1; Met 1-Pro 524).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cellular Markers, Extracellular Matrix, Mesenchymal Stem Cell Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 23769
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.