Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FLRT3 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NB062567 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB062567 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB062567 Supplier Novus Biologicals Supplier No. NBP299728AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FLRT3 Polyclonal specifically detects FLRT3 in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.

Specifications

Antigen FLRT3
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Formulation 50mM Sodium Borate
Gene Alias fibronectin leucine rich transmembrane protein 3, Fibronectin-like domain-containing leucine-rich transmembrane protein 3, HH21, KIAA1469, leucine-rich repeat transmembrane protein FLRT3
Host Species Rabbit
Immunogen Produced in rabbits immunized with purified, recombinant FLRT3 (NP_938205.1; Met 1-Pro 528).
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 23767
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.