Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Antibody (113308), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB346469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB346469 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB346469 Supplier Novus Biologicals Supplier No. FAB7681MFV610
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Mouse samples. It is validated for CyTOF, Flow Cytometry, Immunocytochemistry.

Specifications

Antigen Flt-3/Flk-2/CD135
Applications CyTOF, Flow Cytometry, Immunocytochemistry
Classification Monoclonal
Clone 113308
Conjugate mFluor Violet 610 SE
Dilution Flow Cytometry, Immunocytochemistry, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Host Species Rat
Immunogen Mouse myeloma cell line NS0-derived recombinant mouse Flt-3/Flk-2, Asn28-Ser544 (predicted), Accession # Q00342
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Neuroscience, Neurotransmission, Phospho Specific, Protein Kinase
Primary or Secondary Primary
Gene ID (Entrez) 2322
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.