Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Flt-3/Flk-2/CD135 Antibody (A2F10), Biotin, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP1435820.1MG
Description
Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Flt-3/Flk-2/CD135 | |
Monoclonal | |
0.5 mg/mL | |
Flow Cytometry 1:10-1:1000 | |
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3 | |
Rat | |
Affinity Purified | |
RUO | |
Primary | |
Mouse | |
IgG2a κ |
Flow Cytometry | |
A2F10 | |
Biotin | |
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide | |
FLT3 | |
The immunogen for this antibody was CD135. | |
0.1 mg | |
Cell Cycle and Replication | |
2322 | |
Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction