Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Antibody (A2F10), Biotin, Novus Biologicals™
SDP

Rat Monoclonal Antibody

$610.00 - $987.50

Specifications

Antigen Flt-3/Flk-2/CD135
Clone A2F10
Concentration 0.5 mg/mL
Dilution Flow Cytometry 1:10-1:1000
Applications Flow Cytometry
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP14358201
SDP
View Documents
Novus Biologicals
NBP1435820.1MG
0.1 mg
Each for $610.00
Only null left
Add to Cart
 
NBP1435825
SDP
View Documents
Novus Biologicals
NBP1435820.5MG
0.5 mg
Each for $987.50
Only null left
Add to Cart
 
Description

Description

Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

Flt-3/Flk-2/CD135
0.5 mg/mL
Flow Cytometry
Biotin
RUO
Mouse
CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
FLT3
IgG2a κ
Affinity Purified
A2F10
Flow Cytometry 1:10-1:1000
Monoclonal
Rat
Cell Cycle and Replication
Aqueous buffer, may contain carrier protein/stabilizer (proprietary) with 0.09% Sodium Azide
2322
The immunogen for this antibody was CD135.
Primary
Store at 4C. Do not freeze.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.