Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Antibody (A2F10), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP143352IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP143352IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP143352IR Supplier Novus Biologicals Supplier No. NBP143352IR
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Flt-3/Flk-2/CD135 Monoclonal specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Flow Cytometry.

Specifications

Antigen Flt-3/Flk-2/CD135
Applications Flow Cytometry
Classification Monoclonal
Clone A2F10
Conjugate DyLight 755
Dilution Flow Cytometry
Gene Alias CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Gene Symbols FLT3
Host Species Rat
Immunogen The immunogen for this antibody was CD135.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 2322
Target Species Mouse
Content And Storage Store at 4C in the dark.
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.