Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3/Flk-2/CD135 Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP241188 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP241188 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP241188 Supplier Novus Biologicals Supplier No. NBP241188

Rabbit Polyclonal Antibody

Flt-3/Flk-2/CD135 Polyclonal specifically detects Flt-3/Flk-2/CD135 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen Flt-3/Flk-2/CD135
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 2 ug/ml, ELISA, Immunocytochemistry/Immunofluorescence 20 ug/ml
Formulation PBS with 0.02% Sodium Azide
Gene Alias CD135, CD135 antigen, EC 2.7.10, fetal liver kinase 2, FL cytokine receptor, FLK2, FLT-3, FLT3 receptor tyrosine kinase, Fms-like tyrosine kinase 3, fms-related tyrosine kinase 3, growth factor receptor tyrosine kinase type III, Stem cell tyrosine kinase 1, STK-1, STK1EC 2.7.10.1, Tyrosine-protein kinase receptor FLT3
Gene Symbols FLT3
Host Species Rabbit
Immunogen Antibody was raised against an 18 amino acid synthetic peptide near the center of human FLT3. The immunogen is located within amino acids 340 - 390 of FLT3. Amino Acid Squence: INATNSSEDYEIDQYEEF
Molecular Weight of Antigen 109 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2322
Test Specificity At least three isoforms of FLT3 are known to exist; this antibody will detect all isoforms.
Target Species Human, Mouse
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.