Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Flt-3 Ligand/FLT3L Antibody (008), PE, Novus Biologicals™
SDP

Catalog No. NB307678 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB307678 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB307678 Supplier Novus Biologicals Supplier No. NBP289408PE
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Flt-3 Ligand/FLT3L Monoclonal antibody specifically detects Flt-3 Ligand/FLT3L in Human samples. It is validated for ELISA

Specifications

Antigen Flt-3 Ligand/FLT3L
Applications ELISA
Classification Monoclonal
Clone 008
Conjugate PE
Dilution ELISA
Formulation PBS
Gene Alias FL, Flt3 ligand, FLT3L, fms-related tyrosine kinase 3 ligand, SL cytokine
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human Flt-3 Ligand/FLT3L (Uniprot#: P49771-1; Thr27-Pro185).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Biologically Active Proteins, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 2323
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.