Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fludarabine
GREENER_CHOICE

Catalog No. p-4816731
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Purine analog; inhibits DNA synthesis

Purine analog that inhibits DNA synthesis. Exhibits antiproliferative activity (IC50 = 1.54 μM in RPMI cells) and triggers apoptosis through increasing Bax and decreasing Bid, XIAP and survivin expression. Inhibits cytokine-induced activation of STAT1 and STAT1-dependent gene transcription in lymphocytes. Also displays anticancer activity against hematological malignancies in vivo.

Chemical Identifiers

CAS 21679-14-1
Molecular Formula C10H12FN5O4

Specifications

Quantity 10 mg
Formula Weight 285.23
Purity >99%
Molecular Weight (g/mol) 285.24
Chemical Name or Material Fludarabine
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.