Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescamine, pure

Catalog No. p-4509671
Change view
Click to view available options
Quantity:
25 mg
100 mg
500 mg
2.5 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC191670250 25 mg Glass bottle
AC191671000 100 mg Glass bottle
AC191675000 500 mg Glass bottle
AC191670025 2.5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC191670250 Supplier Thermo Scientific Chemicals Supplier No. 191670250
Only null left
Add to Cart

CAS: 38183-12-9 | C17H10O4 | 278.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 38183-12-9
Molecular Formula C17H10O4
Molecular Weight (g/mol) 278.26
MDL Number MFCD00005928
InChI Key ZFKJVJIDPQDDFY-UHFFFAOYNA-N
PubChem CID 37927
IUPAC Name 4'-phenylspiro[2-benzofuran-3,2'-furan]-1,3'-dione
SMILES O=C1OC2(OC=C(C2=O)C2=CC=CC=C2)C2=CC=CC=C12

Specifications

Melting Point 155.0°C to 157.0°C
Solubility Information Solubility in water: .. Other solubilities: soluble in diethyl ether
Infrared Spectrum Authentic
Formula Weight 278.26
Grade Pure
Packaging Glass bottle
Specific Extinction 900 (235nm, ether) min.
Color Yellow
Quantity 25 mg
Physical Form Powder
Chemical Name or Material Fluorescamine
Show More Show Less
Hazard Category Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.