Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Antibody (4-4-20 (enhanced)), Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB014232 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014232 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB014232 Supplier Novus Biologicals Supplier No. NBP281024APC
Add to Cart
Add to Cart

Human Monoclonal Antibody

Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Flow Cytometry.

Specifications

Antigen Fluorescein
Applications Flow Cytometry
Classification Monoclonal
Clone 4-4-20 (enhanced)
Conjugate APC
Dilution Flow Cytometry
Formulation PBS
Gene Alias FITC, fluorescein isothyocianate
Host Species Human
Immunogen Fluorescein I-bovine serum albumin (FlI-BSA).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Fluorescein labelled proteins.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.