Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Antibody (4-4-20 (enhanced)), DyLight 594, Novus Biologicals™
SDP

Catalog No. NB343257 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB343257 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB343257 Supplier Novus Biologicals Supplier No. NBP252638DL594
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Fluorescein Monoclonal antibody specifically detects Fluorescein in Nonspecies specific samples. It is validated for Western Blot, Flow Cytometry, Control, CyTOF

Specifications

Antigen Fluorescein
Applications Western Blot, Flow Cytometry, Control, CyTOF
Classification Monoclonal
Clone 4-4-20 (enhanced)
Conjugate DyLight 594
Dilution Western Blot, Flow Cytometry, Control, CyTOF-ready
Formulation 50mM Sodium Borate
Gene Alias FITC, fluorescein isothyocianate
Host Species Rabbit
Immunogen Fluorescein I-bovine serum albumin (FlI-BSA).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Non-species specific
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.