Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein Antibody, Texas Red, Novus Biologicals™
SDP

Catalog No. NB001774 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB001774 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB001774 Supplier Novus Biologicals Supplier No. NB1103648100UG
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Fluorescein Polyclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay.

Specifications

Antigen Fluorescein
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen)
Classification Polyclonal
Conjugate Texas Red
Dilution Western Blot >1:10000, Immunohistochemistry, Immunocytochemistry/Immunofluorescence >1:20000, Immunohistochemistry-Frozen, Fluorophore-linked immunosorbent assay >1:20000
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2, 10 mg/mL Bovine Serum Albumin (BSA) - Immunoglobulin and Protease free
Gene Alias FITC, fluorescein isothyocianate
Host Species Goat
Immunogen Fluorescein conjugated to Goat IgG.
Purification Method Affinity Purified
Quantity 100 μg
Test Specificity Fluorescein
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.