Learn More
Fluorescein (FITC) Monoclonal Antibody (8B9.C6.D3), Rockland™

Mouse Monoclonal Antibody
$178.35 - $2702.99
Specifications
| Antigen | Fluorescein (FITC) |
|---|---|
| Clone | 8B9.C6.D3 |
| Concentration | 1 mg/mL |
| Applications | ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot |
| Classification | Monoclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-199-0500
![]()
|
Rockland Immunochemicals
200301096 |
100 μg |
Each for $460.02
|
|
|||||
50-199-0502
![]()
|
Rockland Immunochemicals
200301096S |
25 μL |
Each for $178.35
|
|
|||||
50-199-0501
![]()
|
Rockland Immunochemicals
200301096L |
1 mg |
Each for $2,702.99
|
|
|||||
Description
Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-mouse serum, Fluorescein conjugated IgG and Fluorescein conjugated Bovine Serum Albumin.
FITC (fluorescein isothiocyanate) is a fluorochrome dye that absorbs ultraviolet or blue light causing molecules to become excited and emit a visible yellow-green light. This emission ceases upon removal of the light causing the excitation. Fluorochrome labeling provides rapid, accurate localization of antigen-antibody interaction when one of the reactants is part of a cell, tissue or other biological structure. FITC is a commonly used marker for antibodies in immunofluorescent techniques since the conjugation of FITC to proteins is relatively easy and does not, in general, destroy the biological activity of the labeled protein. FITC is widely used as a hapten to label different proteins. Anti-FITC antibodies can be used for isolating FITC labeled cells by magnetic microbeads based sorting, as well as removal of background signal.Specifications
| Fluorescein (FITC) | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Fluorescein | |
| IgG2a κ | |
| -20°C, Avoid Freeze/Thaw Cycles |
| 8B9.C6.D3 | |
| ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, In Situ Hybridization (ISH), Western Blot | |
| Unconjugated | |
| Mouse | |
| 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2 | |
| Fluorescein conjugated to Mouse IgG | |
| Primary | |
| Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.