Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fluorescein, Rat anti-Non-species specific, Clone: 4-4-20 (enhanced), Novus Biologicals™
SDP

Catalog No. NB014274 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.2 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB014274 0.025 mg
NB014275 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB014274 Supplier Novus Biologicals Supplier No. NBP2810260.025MG
Add to Cart
Add to Cart

Rat Monoclonal Antibody

Fluorescein Monoclonal specifically detects Fluorescein in Non-species specific samples. It is validated for Western Blot, Flow Cytometry, Control.

Specifications

Antigen Fluorescein
Applications Western Blot, Flow Cytometry
Classification Monoclonal
Clone 4-4-20 (enhanced)
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot, Flow Cytometry, Control
Formulation PBS
Gene Alias FITC, fluorescein isothyocianate
Host Species Rat
Immunogen Fluorescein I-bovine serum albumin (FlI-BSA).
Purification Method Protein A purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Fluorescein labelled proteins.
Target Species Non-species specific
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.