Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Agilent BioTek Fluorescence Test Plate for Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Reader
GSA_VAGREENER_CHOICE

Catalog No. bt1400501
Click to view available options
For Use With (Equipment):
Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Readers

Includes

Certificate of calibration, Flash drive with user manual and test protocols, Dust blower and cleaning swabs

Designed for conducting fluorescence intensity, fluorescence polarization, and time-resolved fluorescence qualification tests on Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Readers.

  • Designed to replace fluorescent liquid testing to check instrument performance
  • Solid test plate
  • Immune to the pipetting errors and evaporation issues
  • Convenient size and multi-parameter functionality
  • Checks for fluorescence intensity linearity-green (excitation 485nm, emission 528nm), blue (excitation 360nm, emission 460nm), Carrier flatness, Fluorescence polarization and Time-resolved fluorescence

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type Fluorescence Test Plate
For Use With (Equipment) Synergy Neo2/H1/2/HTX, FLx800 and Cytation Multi-Mode Readers
Excitation/Emission EX 485 nm, EM 528 nm
For Use With (Application) Fluorescence
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.