Description
- For use in lessons on Bowen's reaction series
- Has Mohs scale hardness of 4
- For lessons about hardness and cleavage, as well as the fickle method of identifying minerals by color
- Correlates well to Next Generation Science Standards Evidence Statements
Specifications
Specifications
| Product Type | Mineral |
| For Use With (Application) | Study of uncommon 4-plane cleavage pattern |
| Packaging | Clear labeled and barcoded bag |
| Quantity | 12/Pk. |
| Specimen Size | 2 to 3 cm |
| Specimen Type | Fluorite |