EVOS™ Fluorite Objective Lens
For fluorescence and demanding transmitted-light applications
Manufacturer: EVOS™ AMEP4698
 View more versions of this product
Catalog No. 12-563-586
  Check Availability
Add to Cart
    Specifications
    Product Type Objective lens
    Immersion Type Not Applicable
    Numerical Aperture 0.50
    Magnification Power 20X
    Focus Distance 2.5mm
    Type Plan Fluorite Objective Lens
    For Use With (Equipment) EVOS™ XL Core Imaging System, EVOS FL Color Imaging System, EVOS FL Auto Imaging System, EVOS XL Imaging System, EVOS FL Imaging System
  • Description & Specifications
Related Products
Product Content Feedback