Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Fluoxetine hydrochloride
GREENER_CHOICE

Catalog No. p-4783464
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

5-HT reuptake inhibitor

Fluoxetine hydrochloride is a selective serotonin reuptake inhibitor. Binds to the human 5-HT transporter with a Ki of 0.9 nmol/l and is between 150- and 900-fold selective over 5-HT1A, 5-HT2A, H1, α1, α2-adrenergic, and muscarinic receptors. Antidepressant. Induces differentiation of neuronal precursors, enhancing neuronal characteristics. Fluoxetine also inhibits assembly and activation of the NLRP3-ASC inflammasome and prevents degeneration of retinal pigmented epithelium (RPE) cells in an animal model of dry age-related macular degeneration (AMD).

Chemical Identifiers

CAS 56296-78-7
Molecular Formula C17H19ClF3NO

Specifications

Quantity 10 mg
Formula Weight 345.78
Purity >99%
Molecular Weight (g/mol) 345.79
Chemical Name or Material Fluoxetine hydrochloride
Recommended Storage Store at Room Temperature
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.