Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMNL1 Antibody, DyLight 594, Novus Biologicals™
SDP

Catalog No. NB190238 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB190238 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB190238 Supplier Novus Biologicals Supplier No. NBP281988DL594
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FMNL1 Polyclonal antibody specifically detects FMNL1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunofluorescence

Specifications

Antigen FMNL1
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias C17orf1BMGC133052, C17orf1KW-13, CLL-associated antigen KW-13, FHOD4, FMNLMGC1894, formin-like, formin-like 1, formin-like protein 1, Leukocyte formin, MGC21878
Host Species Rabbit
Immunogen Rabbit polyclonal FMNL1 antibody was raised against a 16 amino acid peptide near the carboxy terminus of human FMNL1. The immunogen is located within the last 50 amino acids of FMNL1. Amino Acid Squence: CTVPFTARTGKRTSRLL
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 752
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.