Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP132469 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP132469 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP132469 Supplier Novus Biologicals Supplier No. NBP132469
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

FMO1 Polyclonal specifically detects FMO1 in Human samples. It is validated for Western Blot.

Specifications

Antigen FMO1
Applications Western Blot
Classification Polyclonal
Concentration 1.0mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal
Gene Alias dimethylaniline monooxygenase [N-oxide-forming] 1, Dimethylaniline oxidase 1, EC 1.14.13.8, Fetal hepatic flavin-containing monooxygenase 1, flavin containing monooxygenase 1, Flavin-containing monooxygenase 1 (fetal liver), FMO 1
Gene Symbols FMO1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human FMO1. The exact sequence is proprietary.
Molecular Weight of Antigen 60 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2326
Test Specificity Tested in human. Not yet tested in any other species.
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.