Learn More
Description
Specifications
Specifications
| Antigen | FMO3 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.91mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunocytochemistry/Immunofluorescence 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000 |
| Formulation | 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal |
| Gene Alias | dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU |
| Gene Symbols | FMO3 |
| Host Species | Rabbit |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
