Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMO3 Antibody (OTI3H1), Alexa Fluor™ 750, Novus Biologicals™
SDP

Catalog No. NP2707A70 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP2707A70 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP2707A70 Supplier Novus Biologicals Supplier No. NBP270755AF750
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FMO3 Monoclonal specifically detects FMO3 in Human samples. It is validated for Western Blot, Immunohistochemistry.

Specifications

Antigen FMO3
Applications Western Blot, Immunohistochemistry
Classification Monoclonal
Clone OTI3H1
Conjugate Alexa Fluor 750
Dilution Western Blot, Immunohistochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias dimethylaniline monooxygenase [N-oxide-forming] 3, Dimethylaniline oxidase 3, dJ127D3.1, EC 1.14.13.8, flavin containing monooxygenase 3, FMO 3, FMO form 2, FMO II, FMOII, Hepatic flavin-containing monooxygenase 3, hepatic flavin-containing monooxygenase-3, MGC34400, TMAU
Gene Symbols FMO3
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-291 of human FMO3 (NP_008825) produced in E.coli.
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 2328
Target Species Human
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.