Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
FMR1 Mouse anti-Mouse,Human,Rat, Unconjugated, Clone: 1F1, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | FMR1 |
---|---|
Clone | 1F1 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABN24532MI
|
MilliporeSigma
MABN245325UG |
25 μg |
Each for $189.29
|
|
|||||
MABN24531MI
|
MilliporeSigma
MABN2453100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
FMR1 Monoclonal antibody specifically detects FMR1 in Mouse,Human,Rat samples. It is validated for Western Blotting.Specifications
FMR1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Mouse, Human, Rat | |
Fmr1, Fmr-1 | |
IgG1 κ | |
Protein G Purified | |
Clone 1F1 is a mouse monoclonal antibody that specifically detects Fragile X mental retardation protein 1 homolog (FMR1). It targets an epitope with in the C-terminal half. |
1F1 | |
Monoclonal | |
Purified | |
Neuroscience | |
Synaptic functional regulator FMR1;Fragile X mental retardation protein 1 homolog;FMRP;Protein FMR-1;mFmr1p | |
Full-length recombinant mouse Fragile X mental retardation protein 1 homolog (FMR1). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼72 kDa observed; 68.99 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title