Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

FMRP Antibody (OTI3B4), PE/Atto594, Novus Biologicals™
SDP

Catalog No. N270761P594 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N270761P594 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N270761P594 Supplier Novus Biologicals Supplier No. NBP270761PEATT594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

FMRP Monoclonal specifically detects FMRP in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen FMRP
Applications Flow Cytometry
Classification Monoclonal
Clone OTI3B4
Conjugate PE/ATTO 594
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias FMRPFRAXAPOF, fragile X mental retardation 1, fragile X mental retardation 1 protein, MGC87458, POF1, premature ovarian failure 1, Protein FMR-1
Gene Symbols FMR1
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids FMR1(NP_002015) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Epigenetics, microRNA
Primary or Secondary Primary
Gene ID (Entrez) 2332.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.