Learn More
FN3KRP Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | FN3KRP |
---|---|
Concentration | 0.24 mg/mL |
Applications | Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-173-0868
![]() ![]() |
Proteintech Group Inc
163121AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
Product No. 16312-1-AP is a rabbit polyclonal antibody raised against the full-length of human FN3KRP. The MW of this protein is 34 kDa, and this antibody specifically recognizes the 34 kDa protein.
FN3KRP and FN3K.Specifications
FN3KRP | |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
Q8K274, Q9HA64 | |
238024, 303755, 79672 | |
FN3KRP Fusion Protein Ag9347 | |
Primary | |
-20°C | |
FN3KRP |
0.24 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
FN3K related protein, FN3K RP, FN3KL, FN3KRP, Ketosamine 3 kinase | |
FN3KRP | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.